Learn More
Cellpath Stainless-steel Reusable Base Molds
Codice prodotto. 30226275
Cambia vista
Click to view available options
Dimensioni (L x P x A):
7 x 7 x 5 mm
10 x 24 x 5 mm
14 x 24 x 5 mm
24 x 24 x 5 mm
30 x 24 x 9 mm
37 x 24 x 9 mm
Dimensione della confezione:
pezzi 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensioni (L x P x A) Dimensione della confezione
30226275 30 x 24 x 9 mm pezzi 5
30226270 7 x 7 x 5 mm pezzi 5
30226276 37 x 24 x 9 mm pezzi 5
30226272 14 x 24 x 5 mm pezzi 5
30226274 24 x 24 x 5 mm pezzi 5
30226271 10 x 24 x 5 mm pezzi 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Codice prodotto. 30226275 Fornitore Cellpath N. del fornitore GBB301405A
  • ONLINE EXCLUSIVE PRICE
    27.22€
    23.20€ / pezzi 5
    Risparmia 4.02€
    14% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 24-11-2026
Ideal for prostrate and breast cores and can be used with both grid and slatted processing cassettes for both horizontal and vertical sectioning. Cellpath Stainless-steel Reusable Base Moulds are manufactured from high quality stainless steel that put an end to extensive trimming of wax.
  • Made of reusable Stainless steel
  • Stackable to reduce storage area
  • Internal rounded corners enable easy removal of the wax block
  • Features extended edges prevent tipping and provide grip area for ease of handling
  • Available in wide range of sizes
  • GBB-0714-05A suitable for use with needle and punch biopsies
Specifica
Materiale Stainless Steel
Dimensioni (L x P x A) 30 x 24 x 9 mm
Quantità 5/Pk.