Learn More
6-Aminoquinolyl-N-hydroxysuccinimidyl Carbamate (90%), TRC
Product Code. 17904365
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17904365 10 mg 10mg
30138244 250 mg 250mg
17991805 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17904365 Supplier Toronto Research Chemicals Supplier No. TRCA62914010MG
 Restricted Item
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 148757-94-2
Molecular Formula C14 H11 N3 O4
Molecular Weight (g/mol) 285.25
Synonym N-6-Quinolinylcarbamic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, 1-[[(6-Quinolinylamino)carbonyl]oxy]-2,5-pyrrolidinedione, AHC Reagent, AccQ.Fluor, AccQTag;
IUPAC Name (2,5-dioxopyrrolidin-1-yl) N-quinolin-6-ylcarbamate
SMILES O=C(Nc1ccc2ncccc2c1)ON3C(=O)CCC3=O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C14H11N3O4/c18-12-5-6-13(19)17(12)21-14(20)16-10-3-4-11-9(8-10)2-1-7-15-11/h1-4,7-8H,5-6H2,(H,16,20)
Formula Weight 285.075
Chemical Name or Material 6-Aminoquinolyl-N-hydroxysuccinimidyl Carbamate