missing translation for 'onlineSavingsMsg'
Learn More

6-chloroimidazo[2,1-b][1,3]thiazole, 97%, Thermo Scientific™

Catalog Number p-2703353
Click to view available options
Quantità:
1 g
5 g
10 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
23576-81-0
C5H3ClN2S
158.603
MFCD00173802
WOZMDYAJHVHPMD-UHFFFAOYSA-N
721582
6-chloroimidazo[2,1-b][1,3]thiazole
C1=CSC2=NC(=CN21)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
23576-81-0
C5H3ClN2S
158.603
MFCD00173802
WOZMDYAJHVHPMD-UHFFFAOYSA-N
721582
6-chloroimidazo[2,1-b][1,3]thiazole
C1=CSC2=NC(=CN21)Cl
Materiale o nome chimico 6-chloroimidazo[2, 1-b][1, 3]thiazole
CAS min. % 97.0
Intervallo percentuale saggio 97%
Imballaggio Amber glass bottle
Formula lineare C6H5CH2CH2NCS
Quantità 1 g
Sinonimo 6-chloroimidazo 2,1-b thiazole, 6-chloroimidazo 2,1-b 1,3 thiazole, 6-chloroimidazole 2,1-b 1,3 thiazole, 6-chloroimidazo 2,1-b 1,3-thiazoline, chloroimidazobthiazole, acmc-1cip3, 6-chloro-imidazo 2,1-b thiazole, imidazo 2,1-b thiazole,6-chloro, imidazo 2,1-b thiazole, 6-chloro, 6-chloro-4h-imidazo 2,1-b thiazole
Peso formulazione 158.61
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'