Learn More
6-Maleimidocaproic Acid Hydrazide, Trifluoroacetic Acid Salt, TRC
Product Code. 30468162
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468162 100 mg 100mg
30466514 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30468162 Supplier Toronto Research Chemicals Supplier No. TRCM136800100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 151038-94-7
Molecular Formula C10 H15 N3 O3 . C2 H F3 O2
Molecular Weight (g/mol) 339.27
IUPAC Name 6-(2,5-dioxopyrrol-1-yl)hexanehydrazide;2,2,2-trifluoroacetic acid
SMILES NNC(=O)CCCCCN1C(=O)C=CC1=O.OC(=O)C(F)(F)F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H15N3O3.C2HF3O2/c11-12-8(14)4-2-1-3-7-13-9(15)5-6-10(13)16;3-2(4,5)1(6)7/h5-6H,1-4,7,11H2,(H,12,14);(H,6,7)
Formula Weight 339.1042
Chemical Name or Material 6-Maleimidocaproic Acid Hydrazide, Trifluoroacetic Acid