Learn More
Alfa Aesar™ 6-Nitroveratraldehyde, 96%
Marca: Alfa Aesar™ A14415.09
Dettagli aggiuntivi : CAS : 20357-25-9 Peso : 0.01000kg
Quantità | 10g |
---|
Descrizione
Applications6-Nitroveratraldehyde has been used in the preparation of no-carrier-added 6-18F-fluoro-L-dopa, fundamental tracer for cerebral positron emission tomography studies of dopaminergic system in humans and o-nitroaryl-bis(5-methylfur-2-yl)methanes, versatile synthons for the synthesis of nitrogen-containing heterocycles. DMNB is also an enzyme involved in the non-homologous end-joining (NHEJ) pathway of double-stranded DNA break (DSB) repair. It is also used for synthesizing analogs of alpha-asarone.
Solubility
Insoluble in water. Soluble to 25 mM in ethanol with gentle warming and to 100 mM in DMSO.
Notes
Air and light sensitive. Store away from air, light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Identificatori chimici
20357-25-9 | |
211.173 | |
YWSPWKXREVSQCA-UHFFFAOYSA-N | |
88505 | |
COC1=C(C=C(C(=C1)C=O)[N+](=O)[O-])OC |
C9H9NO5 | |
MFCD00007134 | |
6-nitroveratraldehyde, dmnb, 3,4-dimethoxy-6-nitrobenzaldehyde, 4,5-dimethoxy-2-nitro-benzaldehyde, dna-pk inhibitor, benzaldehyde, 4,5-dimethoxy-2-nitro, 2-nitro-4,5-dimethoxybenzaldehyde, dna-dependent protein kinase inhibitor, 2-nitro-4,5-bis methyloxy benzaldehyde, pubchem8213 | |
4,5-dimethoxy-2-nitrobenzaldehyde |
Specifica
6-Nitroveratraldehyde | |
131°C to 135°C | |
20357-25-9 | |
MFCD00007134 | |
6-nitroveratraldehyde, dmnb, 3,4-dimethoxy-6-nitrobenzaldehyde, 4,5-dimethoxy-2-nitro-benzaldehyde, dna-pk inhibitor, benzaldehyde, 4,5-dimethoxy-2-nitro, 2-nitro-4,5-dimethoxybenzaldehyde, dna-dependent protein kinase inhibitor, 2-nitro-4,5-bis methyloxy benzaldehyde, pubchem8213 | |
YWSPWKXREVSQCA-UHFFFAOYSA-N | |
4,5-dimethoxy-2-nitrobenzaldehyde | |
88505 | |
96% |
Air and light sensitive | |
10g | |
C9H9NO5 | |
395599 | |
Insoluble in water. Soluble to 25 mM in ethanol with gentle warming and to 100 mM in DMSO. | |
COC1=C(C=C(C(=C1)C=O)[N+](=O)[O-])OC | |
211.173 | |
211.17 |