missing translation for 'onlineSavingsMsg'
Learn More

6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate (6beta-Hydroxymedroxyprogesterone Acetate), Mikromol™

Codice prodotto. 30459124
Cambia vista
Click to view available options
Quantità:
25 mg
100 mg
4 x 25 mg
Dimensione della confezione:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30459124 4 x 25 mg -
30052046 25 mg 25mg
30049568 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30459124 Supplier Mikromol Supplier No. MM0532.0100254AMP

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate (6beta-Hydroxymedroxyprogesterone Acetate), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 6beta-Hydroxymedroxyprogesterone Acetate
CAS 984-47-4
Nomi componenti o analita 6beta-Hydroxymedroxyprogesterone Acetate
Shipping Condition Room Temperature
Formula molecolare C24 H34 O5
Certificazione/Conformità ISO 17025
Sinonimo 6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate, Medroxyprogesterone Acetate Imp. A (EP), 6beta-Hydroxymedroxyprogesterone Acetate, Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-hydroxy-6-methyl-, (6beta)-, Pregn-4-ene-3,20-dione, 6beta,17-dihydroxy-6-methyl-, 17-acetate (7CI,8CI), (6beta)-17-(Acetyloxy)-6-hydroxy-6-methylpregn-4-ene-3,20-dione, Medroxyprogesterone Acetate Impurity A
InChI Formula InChI=1S/C24H34O5/c1-14(25)24(29-15(2)26)11-8-19-17-13-23(5,28)20-12-16(27)6-9-21(20,3)18(17)7-10-22(19,24)4/h12,17-19,28H,6-11,13H2,1-5H3/t17-,18+,19+,21-,22+,23-,24+/m1/s1
Tipo di soluzione Neat
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C[C@@](C)(O)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
IUPAC Name [(6R,8R,9S,10R,13S,14S,17R)-17-acetyl-6-hydroxy-6,10,13-trimethyl-3-oxo-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 402.52
Quantità 4 x 25 mg
Peso formulazione 402.2406 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.