missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(6R)-6-Methyl-octanoic Acid, TRC
Specifications
Specifications
| Materiale o nome chimico | (6R)-6-Methyl-octanoic Acid |
| CAS | 97749-11-6 |
| Formula molecolare | C9H18O2 |
| Quantità | 250 mg |
| Sinonimo | (R)-6-Methyl-octanoic Acid, (6R)-6-Methyloctanoic Acid, (-)-6-Methyloctanoic Acid |
| InChI Formula | InChI=1S/C9H18O2/c1-3-8(2)6-4-5-7-9(10)11/h8H,3-7H2,1-2H3,(H,10,11)/t8-/m1/s1 |
| SMILES | CC[C@@H](C)CCCCC(=O)O |
| IUPAC Name | (6R)-6-methyloctanoic acid |
| Molecular Weight (g/mol) | 158.24 |
| Peso formulazione | 158.1307 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?