Learn More
Univet™ 6X3 Goggles for PPE
Description
- Certified according to EN166 standard, point 3 and 4 for protection against droplets, splashes and large dust particles
- Panoramic lens offers ideal peripheral vision and is easily integrated with prescription glasses
- Fast interchange lens system
- Frame engineered for a soft facial support
- Compatible with filtering half masks
- Lens with anti-fog and anti-scratch ultra performing Vanguard UDC treatment
Specifications
Specifications
| Forma | Rectangular |
| Lens Coating | Anti-fog, Scratch-resistant |
| Lens Tint | None |
| Product Type | Heat-resistant Safety Goggles |
| Materiale | Polycarbonate |
| Monouso | Reusable |
| Lens Material | Policarbonato |
| Materiale del telaio | PCB |
| UV Protection | UV380 |
| Frame Type | Frameless |
| Show More |