missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(6Z)-8-Methyl-6-nonenoic Acid (up to 10% trans isomer), TRC
Specifications
Specifications
| Materiale o nome chimico | (6Z)-8-Methyl-6-nonenoic Acid |
| CAS | 31467-60-4 |
| Formula molecolare | C10 H18 O2 |
| Quantità | 1 g |
| Sinonimo | (Z)-8-Methyl-6-Nonenoic Acid, 8-Methyl-cis-6-nonenoic Acid, cis-8-Methyl-6-nonenoic Acid |
| InChI Formula | InChI=1S/C10H18O2/c1-9(2)7-5-3-4-6-8-10(11)12/h5,7,9H,3-4,6,8H2,1-2H3,(H,11,12)/b7-5- |
| SMILES | CC(C)\C=C/CCCCC(=O)O |
| IUPAC Name | (Z)-8-methylnon-6-enoic acid |
| Molecular Weight (g/mol) | 170.25 |
| Peso formulazione | 170.1307 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?