Learn More
7-Dehydrocholesterol Oleate, TRC
Product Code. 30368059
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368059 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30368059 Supplier Toronto Research Chemicals Supplier No. TRCD229475100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 7-Dehydrocholesterol Oleate
CAS 10105-12-1
Molecular Formula C45H76O2
Quantity 100 mg
Synonym Cholesta-5,7-dien-3-ol, (9Z)-9-octadecenoate, (3beta)- (9CI), Cholesta-5,7-dien-3-ol, 9-octadecenoate, [3beta(Z)]-
InChI Formula InChI=1S/C45H76O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-25-43(46)47-38-30-32-44(5)37(34-38)26-27-39-41-29-28-40(36(4)24-22-23-35(2)3)45(41,6)33-31-42(39)44/h14-15,26-27,35-36,38,40-42H,7-13,16-25,28-34H2,1-6H3/b15-14-/t36-,38+,40-,41+,42+,44+,45-/m1/s1
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4C3=CC=C2C1)[C@H](C)CCCC(C)C
IUPAC Name [(3S,9S,10R,13R,14R,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-yl] (Z)-octadec-9-enoate
Molecular Weight (g/mol) 649.08
Formula Weight 648.5845