Learn More
R&D Systems™ 740 Y-P
Product Code. 18776376
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18776376 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18776376 Supplier R&D Systems™ Supplier No. 1983/1
  • ONLINE EXCLUSIVE PRICE
    559.91€
    501.90€ / 1mg
    Risparmia 58.01€
    10% Sconto
 Restricted Item
In stock
Add to basket
Edge
Add to basket
A Tocris product, part of R&D Systems. Cell-permeable PI 3-kinase activator
740 Y-P is a cell-permeable phosphopeptide activator of PI 3-kinase. Binds with high affinity to p85 subunit of the enzyme. Displays mitogenic activity in C2 muscle cells and promotes survival of rat cerebellar granule neurons in vitro.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Tocris
Quantity 1 mg