Learn More
8-Azaadenine, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Azaadenine |
| CAS | 1123-54-2 |
| Molecular Formula | C4H4N6 |
| Quantity | 50 mg |
| Synonym | 1H-1,2,3-Triazolo[4,5-d]pyrimidin-7-amine, 6-Amino-8-azapurine, 7-Amino-1-v-triazolo[d]pyrimidine, 7-Amino-v-triazolo[d]pyrimidine, 8-Aza-6-aminopurine, NSC 32797 |
| InChI Formula | InChI=1S/C4H4N6/c5-3-2-4(7-1-6-3)9-10-8-2/h1H,(H3,5,6,7,8,9,10) |
| SMILES | NC(NC=N1)=C2C1=NN=N2 |
| IUPAC Name | 1H-triazolo[4,5-d]pyrimidin-7-amine |
| Molecular Weight (g/mol) | 136.11 |
| Formula Weight | 136.0497 |