missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoadenine, Thermo Scientific Chemicals

Catalog Number p-7043111
Click to view available options
Quantità:
100 mg
500 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6974-78-3
C5H4BrN5
214.03
MFCD00082518
FVXHPCVBOXMRJP-UHFFFAOYSA-N
8-bromoadenine, 8-bromo-9h-purin-6-amine, 6-amino-8-bromopurine, 8-br-adenine, 8-bromo-1h-adenine, 8-bromo-1h-purin-6-amine, 8-bromo-9h-purin-6-ylamine, 8-bromopurine-6-ylamine, chembl65747, 9h-purin-6-amine, 8-bromo
81457
8-bromo-7H-purin-6-amine
NC1=C2NC(Br)=NC2=NC=N1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
6974-78-3
C5H4BrN5
214.03
MFCD00082518
FVXHPCVBOXMRJP-UHFFFAOYSA-N
8-bromoadenine, 8-bromo-9h-purin-6-amine, 6-amino-8-bromopurine, 8-br-adenine, 8-bromo-1h-adenine, 8-bromo-1h-purin-6-amine, 8-bromo-9h-purin-6-ylamine, 8-bromopurine-6-ylamine, chembl65747, 9h-purin-6-amine, 8-bromo
81457
8-bromo-7H-purin-6-amine
NC1=C2NC(Br)=NC2=NC=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico 8-Bromoadenine
Punto di fusione >250°C
Colore Brown to White
Quantità 100 mg
Peso formulazione 214.03
Forma fisica Powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'