Learn More
Labconco™ 8-Port Manifold for FreeZone™ Freeze Dryers
Product Code. 15537559 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
1 L, 2.5 L, 4.5 L, 6 L, 8 L, 12 L, or 18 L FreeZone™ Freeze Dryers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15537559 1 L, 2.5 L, 4.5 L, 6 L, 8 L, 12 L, or 18 L FreeZone™ Freeze Dryers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15537559 Supplier Labconco™ Supplier No. 7201800
  • ONLINE EXCLUSIVE PRICE
    2545.00€
    2071.41€ / pezzi 1
    Risparmia 473.59€
    18% Sconto
View all promotions
 Restricted Item
Spedizione stimata: 05-01-2027
Stainless steel manifold for use with FreeZone™ Freeze Dryers includes neoprene valves with plastic knobs.
8-Port Manifold
  • Type 304 stainless steel
  • 8 neoprene valves with plastic knobs accommodate both 1/2 in. and 3/4 in. adapters
  • For use with 1 L, 2.5L, 4.5 L, 6L, 8L, 12 L, or 18 L FreeZone Freeze Dryers
  • Freeze Dryer, glassware, and adapters are required (not included)
  • Includes one latching pin
Specifications
Type Drying Manifold
No. of Shelves 0
Material Stainless Steel (304)
No. of Ports 8
For Use With (Equipment) 1 L, 2.5 L, 4.5 L, 6 L, 8 L, 12 L, or 18 L FreeZone™ Freeze Dryers
Includes 8 neoprene valves with molded plastic knobs and 1 latching pin
Width (English) 8.6 in.
Height (English) 15.7 in.
Length (English) 8.6 in.
Shipping Weight (English) 8.5 lb.
Weight (English) 7 lb.