Learn More
Hamilton™ 800 Microliter Syringes, No Needle Included
Descrizione
- Ideal for dispensing volumes from 0.5 μL up to 250 μL
- A variety of different terminations and needle options are available
- Syringe Holder is attached to the flange that prevents the barrel from heating up and provides support to small volume plungers that are prone to bending
Specifica
Specifica
| Materiale | Borosilicate Glass |
| Calibro dell'ago | No Needle Included |
| Product Type | Microliter Syringe |
| Tipo di collegamento | Removable |
| Graduated | Yes |