Learn More
Thermo Scientific™ 8000 Series Silica Particle Size Standards
Codice prodotto. 11884143 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Diametro (sistema metrico):
0.49μm ± 0.02μm
0.7 μm
1 μm
1.5 μm
Dimensione della confezione:
15mL
Pezzo
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Diametro (sistema metrico) unitSize
11884143 0.7 μm 15mL
11503153 1 μm Pezzo
11894143 1 μm 15mL
11884153 1.5 μm 15mL
11874143 0.49μm ± 0.02μm 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Codice prodotto. 11884143 Fornitore Thermo Scientific™ N. del fornitore 8070
  • ONLINE EXCLUSIVE PRICE
    303.00€
    294.80€ / 15mL
    Risparmia 8.20€
    2% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 28-08-2026
Aggiungi al carrello
Aggiungi al carrello
Provide greater durability and chemical stability when using solvents with these monodisperse silica microspheres used in instrument calibration and as spacers.
Provide greater durability and chemical stability when using solvents with Thermo Scientific™ 8000 Series Silica Particle Size Standards, monodisperse silica microspheres used in instrument calibration and as spacers. Standards are calibrated and certified with NIST™ traceable mean diameters and are suitable for a wide variety of particle measurement applications.
Features:
  • Standards are designed for applications requiring monodisperse inorganic spheres.
  • Opaque spheres provide more contrast than polymer particles in optical and electron beams; like glass, silica has a much higher density that polystyrene.
TRUSTED_SUSTAINABILITY
Specifica
Concentrazione 2% solids
Additivo None
Descrizione Designed for applications requiring monodisperse inorganic spheres, the 8000 Series Silica Particles have a much higher density then the polymer particles.
Diametro (sistema metrico) 0.7 μm
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer. Product comes with stir bar for continual mixing duriong sampling.
Densità particelle 1.8 to 2.2g/cm³
Materiale Amorphous Silica
Include Certificate of Calibration and Traceability to NIST
Stabilità Product stable for a minimum of 24 months
Quantità 15 mL
Indice di rifrazione 1.40 to 1.46 at 589nm (25°C)
Distribuzione dimensionale s = 0.03µm, 4.1% CV
Da utilizzare con (applicazione) Instrument Calibration
Product Type Silica Particle Size Standard