missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-10-phenylanthracene, 98%, Thermo Scientific Chemicals

Catalog Number p-7056052
Click to view available options
Quantità:
250 mg
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
23674-20-6
C20H13Br
333.228
MFCD00230983
WHGGVVHVBFMGSG-UHFFFAOYSA-N
10-bromo-9-phenylanthracene, anthracene, 9-bromo-10-phenyl, 9-bromo-10-phenyl-anthracene, ksc201k3d, 9-bromo-10-phenyl anthracene
4155836
9-bromo-10-phenylanthracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)Br
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
23674-20-6
C20H13Br
333.228
MFCD00230983
WHGGVVHVBFMGSG-UHFFFAOYSA-N
10-bromo-9-phenylanthracene, anthracene, 9-bromo-10-phenyl, 9-bromo-10-phenyl-anthracene, ksc201k3d, 9-bromo-10-phenyl anthracene
4155836
9-bromo-10-phenylanthracene
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(C4=CC=CC=C42)Br

It is used as an OLED material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED material.

Solubility
Insoluble in water. Soluble in hot toluene, dimethylformamide.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 9-Bromo-10-phenylanthracene
Punto di fusione 154°C to 158°C
Colore Yellow
Quantità 250 mg
Beilstein 2277064
Informazioni di solubilità Insoluble in water. Soluble in hot toluene,dimethylformamide.
Peso formulazione 333.23
Percent Purity 98%
Forma fisica Powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'