missing translation for 'onlineSavingsMsg'
Learn More

9-Methyl-3-methylene-1,2,3,9-tetrahydro-4H-carbazol-4-one, Mikromol™

Product Code. 18010510
Change view
Click to view available options
Quantità:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
18010510 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18010510 Supplier Mikromol Supplier No. MM0252.040025

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

9-Methyl-3-methylene-1,2,3,9-tetrahydro-4H-carbazol-4-one
TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 9-Methyl-3-methylene-1,2,3,9-tetrahydro-4H-carbazol-4-one
CAS 99614-64-9
Nomi componenti o analita 9-Methyl-3-methylene-1,2,3,9-tetrahydro-4H-carbazol-4-one
Shipping Condition Room Temperature
Formula molecolare C14 H13 N O
Certificazione/Conformità ISO 17025
Sinonimo Ondansetron Hydrochloride Dihydrate Imp. D (EP), Ondansetron Imp. D (EP), Ondansetron USP Related Compound D, Ondansetron USP RC D, 9-Methyl-3-methylene-1,2,3,9-tetrahydro-4H-carbazol-4-one, GR 44122X, Ph Eur Ondansetron Impurity D, USP Ondanestron Related Compound D, ondansetron olefin (GR 44122X), Ondansetron related, Ondansetron Related Compound D, Ondansetron Hydrochloride Dihydrate Impurity D, Ondansetron Impurity D
InChI Formula InChI=1S/C14H13NO/c1-9-7-8-12-13(14(9)16)10-5-3-4-6-11(10)15(12)2/h3-6H,1,7-8H2,2H3
SMILES Cn1c2CCC(=C)C(=O)c2c3ccccc13
IUPAC Name 9-methyl-3-methylidene-1,2-dihydrocarbazol-4-one
Molecular Weight (g/mol) 211.26
Quantità 25 mg
Peso formulazione 211.0997 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.