Learn More
9-Methylacridine, 97%
Product Code. 10174540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10174540 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10174540 Supplier Thermo Scientific Acros Supplier No. 276490010
 Restricted Item
In stock
Add to basket
CAS: 611-64-3 | C14H11N | 193.25 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 611-64-3
CAS Min % 99.0
CAS Max % 100.0
Melting Point 114.0°C to 119.0°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C14H11N
MDL Number MFCD00143523
Quantity 1 g
Synonym acridine, 9-methyl, 5-methylacridine, 9-methylakridin, 9-methylakridin czech, unii-1f3q4u9uis, 9-methyl acridine, 5-methylacridine european, 1f3q4u9uis, 9-methylacridin, 9-methyl-acridine
InChI Key FLDRLXJNISEWNZ-UHFFFAOYSA-N
SMILES CC1=C2C=CC=CC2=NC2=CC=CC=C12
IUPAC Name 9-methylacridine
Molecular Weight (g/mol) 193.25
PubChem CID 11913
Formula Weight 193.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 9-Methylacridine, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 9-Methylacridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.