Descrizione
- Spectacles combine an attractive appearance with robust quality
- Light, sporty design with panoramic lenses ensure maximum comfort
- Permanent anti-fog performance, even after repeated cleaning
- Soft, duo-component arms for a comfortable non-slip grip
- Flat-end arm design with opening to attach a neck cord
- Available with a sunglare filter (9194885)
Supravision Excellence Coating
- Interior: Anti-fog
- Exterior: Scratch-resistant and chemical-resistant
- Lenses are easy to clean and less susceptible to soiling
Specifica
Specifica
| Certificazione/Conformità | EN 166:2001, EN 172:1994 + A1:2000 + A2:2001 |
| Tipo di lente | Panoramic |
| Lens Coating | Anti-fog |
| Tipo di filtro | Sunglare |
| Lens Tint | Silver Mirror |
| Product Type | Ergonomic Safety Glasses |
| Colore telaio | Black, Gray |
| Modello | 9194 i-Works |
| Valore di trasmittanza | 0.53 |
| Materiale | Polycarbonate |
| Vedi altri risultati |