Descrizione
- Cleverly designed ventilation system helps prevent fogging
- Design is particularly well-suited for use with a respirator
- Wide-vision goggles can be worn comfortably over prescription glasses
- Certified in line with EN 166 (personal eye protection) and EN 170 (UV filters)
- Supravision excellence coating is extremely scratch-resistant and chemical-resistant on the outside, and anti-fog on the inside
- Lenses can be replaced for extended goggle life
Specifica
Specifica
| Tipo di lente | Replaceable, with two tear-off films |
| Lens Coating | Anti-fog |
| Certifications/Compliance (Europe) | EN 166:2001, EN 170:2002 |
| Lens Tint | Clear |
| Product Type | Anti-fog Goggles |
| Colore telaio | Transparent Gray |
| Modello | 9301 Ultravision (Wide-Vision) |
| Materiale | Polycarbonate |
| Monouso | Reusable |
| Lens Material | Policarbonato |
| Vedi altri risultati |