Learn More
Uvex 94909 Comfortable Climate Insole For Uvex G2 - Width 12
Comfortable climatic insole with antistatic properties for Uvex 1 G2
Marca: Uvex 9490942
Dettagli aggiuntivi : Peso : 0.05500kg
Descrizione
- Full-surface foot shock absorption
- Highly breathable, moisture-absorbent and quickly re-drying material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No: 9490.9
- Sizes: 35 to 52
Specifica
Black | |
Microvelour | |
No | |
Wide width | |
No |
Uvex G2 | |
42 | |
Unisex | |
Yes |