Learn More
Uvex 95286 High Tune-Up Insole For Uvex 1 Sport - Width 11
Tune-Up antistatic insole for reducing fatigue and supporting arch
Marca:  Uvex 9528636
Dettagli aggiuntivi : Peso : 0.09400kg
Codice prodotto. 17228091
  • 20.20€ / 1 paio
Spedizione stimata: 26-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Uvex Tune-Up Comfortable High Insole 95286
  • Antistatic insole concept for use in safety footwear
  • Designed to reduce fatigue by increasing wearer comfort and enhancing ergonomics by optimum support of arch of feet
  • Good cushioning
  • Relief of pressure marks
  • Non-irritating, comfortable surface structure
  • Breathable and absorbs moisture
  • Antibacterial
  • Antistatic
  • Useable for ESD-footwear
  • Uvex 1 Sport
  • Article No. 95286
  • Standard EN ISO 20345:2011
  • Sizes: 35 to 52
Specifica
Specifica
Orange, Black
Microvelour
No
Standard width
No
Uvex 1 Sport
36
Unisex