Learn More
TPP™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Descrizione
Color: Clear; Sterility: Sterile; Lid: With Lid
- Opto-mechanical treatment of growth area for optimal cell adhesion
- Lid with air-venting system guarantees controlled gas exchange with low evaporation
- Sloped edge allows placement of lid in one position only
- Yellow marking area on lid and plate ensure correct lid orientation
- Ridged grip area ensures secure grasp and prevents from accidentally lifting off lid
- Clear alpha-numerical labelling between wells for easy orientation during microscopy
- Growth area on spherical zone only
- Absolutely flat growth surface
- Crystal clear transparency
Specifica
Specifica
| Certificazione/Conformità | ANSI/SLAS 1-2004 through ANSI/SLAS 4-2004 |
| Coperchio | With Lid |
| Area di coltura | 0.636 cm2 |
| Imballaggio | 1/Pk., 162/Cs. |
| Colore | Clear |
| Materiale | Polystyrene |
| No. of Wells | 96 |
| Sterilità | Sterile |
| Trattamento superficiale | Cell Culture |
| Total Volume (Metric) Well | 0.34 mL |
| Vedi altri risultati |