Learn More
Uvex™ 9770 Pheos E-WR Unventilated Safety Helmets
Lightweight, unventilated helmets in a sporty design ideal for use in the electrician sector with low voltage systems. Features interior equipment with a rotating wheel system for strapless width adjustment.
Marca:  Uvex™ 9770430
Dettagli aggiuntivi : Peso : 0.40200kg
Codice prodotto. 17299101
  • 18.10€ / Pezzo
Spedizione stimata: 08-03-2023
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Lightweight, unventilated helmet in a sporty design
  • Completely closed for use in the electrician sector with low voltage systems
  • Includes an adapter system for visors, helmet caps, and helmet lamps
  • Interior equipment with a rotating wheel system for stepless width adjustment
  • Reflective helmets are supplied with Reflex Set L pre-attached to increase worker visibility and safety
Specifica
Specifica
EN 397:2012, EN 50365:2002
Non-reflective
Adapter System for Uvex™ Visors, Helmet Capsules, and Helmet Lamps
9770 Pheos E-WR
52 to 61 cm
Green
Low Voltage Systems
HDPE
Electrically Insulating Helmet
Not Ventilated