Learn More
Uvex™ 9790 Winter Caps
The winter cap fits inside all Uvex™ work safety helmets to keep workers warm and comfortable in all types of conditions. Outer material is reflective for increased visibility, in addition to being resistant to wind and water.
32.50€
Specifica
Product Type | Winter Cap |
---|---|
Size | Universal |
Materiale | Fleece-lined Polyester |
Monouso | Reusable |
Da utilizzare con (apparecchiatura) | Uvex™ Safety Helmets |
Codice del prodotto | Marca | Prezzo | Quantità e disponibilità | ||||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Prezzo | Quantità e disponibilità | ||||||
17269201
|
Uvex™
9790068 |
Orange |
32.50€
Pezzo |
Spedizione stimata: 29-05-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
17239201
|
Uvex™
9790065 |
32.50€
Pezzo |
Spedizione stimata: 29-05-2024
Eseguire il login per visualizzare lo stock disponibile |
||||||
Descrizione
- Outer material is windproof and waterproof
- Side mesh increases attention to environmental influences
- Fleece lining is warm and comfortable
- Fluorescent colors increase worker visibility and safety
- Compatible with all Uvex™ Safety Helmets
Specifica
Winter Cap | |
Fleece-lined Polyester | |
Uvex™ Safety Helmets |
Universal | |
Reusable | |
Wind, Water |