Learn More
A 54556A (~90%), TRC
Codice prodotto. 30324168
Cambia vista
Click to view available options
Quantità:
2.5 mg
Dimensione della confezione:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30324168 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30324168 Fornitore Toronto Research Chemicals N. del fornitore TRCA1010552.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico A 54556A
CAS 95398-45-1
Formula molecolare C38H50N6O8
Quantità 2.5 mg
Sinonimo (4R)-N-[(2E,4E,6E)-1-oxo-2,4,6-Octatrien-1-yl]-L-phenylalanyl-L-seryl-L-prolyl-N-methyl-L-alanyl-L-alanyl-4-methyl-L-proline (6→2)-lactone, A 54556 factor A, ADEP 1, Acyldepsipeptide 1, Acyldepsipeptide A, Antibiotic A 54556A
InChI Formula InChI=1S/C38H50N6O8/c1-6-7-8-9-13-18-32(45)40-28(21-27-15-11-10-12-16-27)34(47)41-29-23-52-38(51)31-20-24(2)22-44(31)35(48)25(3)39-33(46)26(4)42(5)37(50)30-17-14-19-43(30)36(29)49/h6-13,15-16,18,24-26,28-31H,14,17,19-23H2,1-5H3,(H,39,46)(H,40,45)(H,41,47)/b7-6+,9-8+,18-13+/t24-,25+,26+,28?,29?,30+,31+/m1/s1
SMILES O=C(C(CC1=CC=CC=C1)NC(/C=C/C=C/C=C/C)=O)NC(COC([C@@H]2C[C@@H](C)CN2C3=O)=O)C(N4CCC[C@H]4C(N(C)[C@@H](C)C(N[C@H]3C)=O)=O)=O
IUPAC Name (2E,4E,6E)-N-[(2S)-1-oxo-3-phenyl-1-[[(3S,7S,9R,13S,16S,19S)-9,13,16,17-tetramethyl-2,6,12,15,18-pentaoxo-5-oxa-1,11,14,17-tetrazatricyclo[17.3.0.07,11]docosan-3-yl]amino]propan-2-yl]octa-2,4,6-trienamide
Molecular Weight (g/mol) 718.84
Peso formulazione 718.369