Learn More
A 54556A (~90%), TRC
Codice prodotto. 30327648
Cambia vista
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30327648 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30327648 Fornitore Toronto Research Chemicals N. del fornitore TRCA1010551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico A 54556A
CAS 95398-45-1
Formula molecolare C38H50N6O8
Quantità 1 mg
Sinonimo (4R)-N-[(2E,4E,6E)-1-oxo-2,4,6-Octatrien-1-yl]-L-phenylalanyl-L-seryl-L-prolyl-N-methyl-L-alanyl-L-alanyl-4-methyl-L-proline (6→2)-lactone, A 54556 factor A, ADEP 1, Acyldepsipeptide 1, Acyldepsipeptide A, Antibiotic A 54556A
InChI Formula InChI=1S/C38H50N6O8/c1-6-7-8-9-13-18-32(45)40-28(21-27-15-11-10-12-16-27)34(47)41-29-23-52-38(51)31-20-24(2)22-44(31)35(48)25(3)39-33(46)26(4)42(5)37(50)30-17-14-19-43(30)36(29)49/h6-13,15-16,18,24-26,28-31H,14,17,19-23H2,1-5H3,(H,39,46)(H,40,45)(H,41,47)/b7-6+,9-8+,18-13+/t24-,25+,26+,28?,29?,30+,31+/m1/s1
SMILES O=C(C(CC1=CC=CC=C1)NC(/C=C/C=C/C=C/C)=O)NC(COC([C@@H]2C[C@@H](C)CN2C3=O)=O)C(N4CCC[C@H]4C(N(C)[C@@H](C)C(N[C@H]3C)=O)=O)=O
IUPAC Name (2E,4E,6E)-N-[(2S)-1-oxo-3-phenyl-1-[[(3S,7S,9R,13S,16S,19S)-9,13,16,17-tetramethyl-2,6,12,15,18-pentaoxo-5-oxa-1,11,14,17-tetrazatricyclo[17.3.0.07,11]docosan-3-yl]amino]propan-2-yl]octa-2,4,6-trienamide
Molecular Weight (g/mol) 718.84
Peso formulazione 718.369