Learn More
a-Amylcinnamaldehyde stabilized with BHT, TRC
Specifica
Specifica
| Materiale o nome chimico | Amylcinnamic aldehyde |
| CAS | 122-40-7 |
| Formula molecolare | C14 H18 O |
| Quantità | 100 mL |
| Sinonimo | Cinnamaldehyde, α-amyl- (4CI), Cinnamaldehyde, α-pentyl- (6CI,7CI,8CI), 2-(Phenylmethylene)heptanal, 2-Benzylideneheptanal, Amyl cinnamal, Amylcinnamaldehyde, Amylcinnamic acid aldehyde, Amylcinnamic aldehyde, Flomine, Jasminal, Jasminaldehyde, Jasmine aldehyde, NSC 6649, Pentylcinnamaldehyde, α-Amyl-β-phenylacrolein, α-Amylcinnamal, α-Amylcinnamaldehyde, α-Pentylcinnamaldehyde |
| InChI Formula | InChI=1S/C14H18O/c1-2-3-5-10-14(12-15)11-13-8-6-4-7-9-13/h4,6-9,11-12H,2-3,5,10H2,1H3/b14-11+ |
| SMILES | CCCCC\C(=C/c1ccccc1)\C=O |
| IUPAC Name | (2E)-2-benzylideneheptanal |
| Molecular Weight (g/mol) | 202.29 |
| Peso formulazione | 202.1358 |