Learn More
a-Methylaminovalerophenone Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Pentedrone (hydrochloride) |
| CAS | 879669-95-1 |
| Formula molecolare | C12 H17 N O . Cl H |
| Quantità | 50 mg |
| Sinonimo | beta-Ethylmethcathinone Hydrochloride, Pentedrone Hydrochloride, a-Methylaminovalerophenone Hydrochloride, 2-(Methylamino)-1-phenyl-1-pentanone Hydrochloride, Pentedrone Hydrochloride |
| InChI Formula | InChI=1S/C12H17NO.ClH/c1-3-7-11(13-2)12(14)10-8-5-4-6-9-10;/h4-6,8-9,11,13H,3,7H2,1-2H3;1H |
| SMILES | Cl.CCCC(NC)C(=O)c1ccccc1 |
| IUPAC Name | 2-(methylamino)-1-phenylpentan-1-one;hydrochloride |
| Molecular Weight (g/mol) | 227.73 |
| Peso formulazione | 227.1077 |