Learn More
ABCA3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA552478
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: MLRLTLGEYG RTVVPFSVPG TSQLGQQLSE HLKDALQAEG QEPREVLGDL EEFLIFRASV EGGGFNERCL VAASFRDVGE RTVVNALFNN QAYHSPATAL AVVDNLLFKL LCGPHASIVV SNFPQPRSAL QAAKDQFNEG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
ABCA3 Polyclonal antibody specifically detects ABCA3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifica
ABCA3 | |
Polyclonal | |
Unconjugated | |
ABCA3 | |
1810036E22Rik; ABC transporter 3; ABC3; ABCA3; ABC-C; ABC-C transporter; ATP binding cassette subfamily A member 3; ATP-binding cassette 3; ATP-binding cassette sub-family A member 3; ATP-binding cassette transporter 3; ATP-binding cassette, subfamily A (ABC1), member 3; ATP-binding cassette, sub-family A (ABC1), member 3; EST111653; LBM180; SMDP3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99758 | |
ABCA3 | |
Recombinant protein corresponding to Human ABCA3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only