missing translation for 'onlineSavingsMsg'
Learn More

Abcam Human Superoxide Dismutase 1 Antibody Pair - BSA and Azide free, 10 x 96 Tests

Codice prodotto. 32113517 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
10 x 96 Tests
Dimensione della confezione:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32113517 10 x 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 32113517 Fornitore Abcam N. del fornitore ab25386610x96Tests
 Articolo soggetto a restrizioni
Spedizione stimata: 03-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Antibody, suitable for sELISA.

  • Applications: sELISA
  • Species Reactivity: Human

Specifications

Applicazioni Sandwich ELISA
Coniugato Unconjugated
Quantità 10 x 96 Tests
Status giuridico RUO
Molecola intera Yes
Primario o secondario Secondary
Test di specificità This product reacts with Human.
Target Species Human
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv sELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.