missing translation for 'onlineSavingsMsg'
Learn More

Abcam Rabbit Anti-Goat IgM H&L (HRP), 1 mg

Codice prodotto. 32117039 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32117039 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 32117039 Fornitore Abcam N. del fornitore ab1128621mg
 Articolo soggetto a restrizioni
Spedizione stimata: 27-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Anti-Goat IgM H&L (HRP)

  • Host Species: Rabbit
  • Clonality: Polyclonal
  • Isotype: IgG
  • Applications: ELISA, ICC/IF, IHC-P, WB
  • Species Reactivity: Goat
  • Storage Buffer: pH: 6.8 - 7.4, Preservative: 0.05% CMIT/MIT based preservative, Constituents: 0.2% BSA
  • Form: Liquid
  • Purification: Affinity purification Immunogen

Specifica

Applicazioni ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classificazione Polyclonal
Coniugato HRP
Formulazione pH: 6.8 - 7.4, Preservative: 0.05% CMIT/MIT based preservative, Constituents: 0.2% BSA
Specie ospite Rabbit
Metodo di purificazione Affinity purification Immunogen
Quantità 1 mg
Status giuridico RUO
Molecola intera Yes
Primario o secondario Secondary
Imballaggio Vial
Test di specificità This product reacts with Goat.
Target Species Goat
Product Type Polyclonal Secondary Antibody
Forma Liquid
Isotype IgG
Host abrv Rb
Shipping Condition Blue Ice
Reactivity abrv Gt
Validated Application abrv ELISA, ICC/IF, IHC-P, WB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.