missing translation for 'onlineSavingsMsg'
Learn More

ABCF2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA554143

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15712081

  • 477.00€ / 100µL
Spedizione stimata: 10-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: KEAAKARQRP RKGHEENGDV VTEPQVAEKN EANGRETTEV DLLTKELEDF EMKKAAARAV TGVLASHPNS TDVHIINLSL TFHGQELLSD TKLELNSGRR YGLIGLNGIG KS Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 97%.

ABCF2 Polyclonal antibody specifically detects ABCF2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifica

Specifica

ABCF2
Polyclonal
Unconjugated
Abcf2
0710005O05Rik; ABC28; Abcf2; ABC-type transport protein; ATP binding cassette subfamily F member 2; ATP-binding cassette sub-family F member 2; ATP-binding cassette, subfamily F (GCN20), member 2; ATP-binding cassette, sub-family F (GCN20), member 2; developmentally regulated repeat element-containing transcript 3; DKFZp586K1823; Drr3; E430001O06; EST133090; HUSSY18; HUSSY-18; Iron-inhibited ABC transporter 2; M-ABC1
Rabbit
Antigen affinity chromatography
RUO
10061, 27407, 311959
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q99LE6, Q9UG63
Abcf2
Recombinant protein corresponding to Human ABCF2
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only