missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ADIPOQ (Human) ELISA Kit

For detection of ADIPOQ in Human Urine and Murine, Adiponectin in Plasma, Serum and cell culture supernatants

Marca:  Abnova™ KA3019.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057765

  • 578.00€ / 1 set


Spedizione stimata
23-10-2020
Aggiungi al carrello

Specifica

Specifica

1.56 to 100ng/mL
Human
10pg/mL
Quantification
ADIPOQ (Human) ELISA Kit
RUO
Store at 4°C for 4 months, or at -20°C for 8 months.
100μL
Biotin
ADIPOQ (Human) ELISA Kit is intended for the quantitative measurement of human ADIPOQ.
Colorimetric
96-well plate pre-coated with anti-Human Adiponectin antibody, Lyophilized Human Adiponectin standards (100ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Human Adiponectin antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated) (130uL), ABC diluent buffer (12mL),TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X)(30mL)
96 Assays
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
96 Assays
Videos
Documenti