missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Cp (Rat) ELISA Kit

Competitive Enzyme Immunoassay for the quantitative measurement of Rat Cp in Plasma and Serum

Marca:  Abnova™ KA2502.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057335

  • 578.00€ / 1 set


Spedizione stimata
29-10-2020
Aggiungi al carrello

Specifica

Specifica

HRP
Cp (Rat) ELISA Kit is for the quantitative measurement of Cp in plasma and serum.
Colorimetric
96 wells Anti mouse ceruloplasmin coated plate, Lyophilized ceruloplasmin stock, 10x Diluent (25mL), Enzyme conjugate reagent (11mL), 20X Wash Solution (50mL),TMB Reagent (11mL), Stop Solution (11mL)
1 Kit
Plasma, Serum
96 Assays
Rat
4.7 to 300ng/mL
Quantification
Cp (Rat) ELISA Kit
RUO
Store the kit at 4°C.
100μL
Videos
Documenti