missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Epstein Barr Virus VCA IgG ELISA Kit

Solid phase Enzyme Immunoassay for the quantitative measurement of IgG class autoantibodies against Epstein Barr Virus VCA

Marca:  Abnova™ KA1444.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16056645

  • 263.00€ / 1 set


Spedizione stimata
21-10-2020
Aggiungi al carrello

Specifica

Specifica

HRP
Colorimetric
96 ELISA break away strip coated with specific Antigen, 1.3mL standard A to F, 15mL Antibodies-Px Conjugate, 125mL Wash Buffer concentrate, 125mL Dilution Buffer, 15mL chromogenic Substrate, 30mL stop solution
Epstein Barr Virus VCA IgG ELISA kit
For research use only (RUO)
Store at 4°C on dry atmosphere away from light.
100μL
Epstein Barr Virus VCA IgG ELISA Kit is a solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Epstein Barr Virus VCA.
Quantification
Epstein Barr Virus VCA IgG ELISA Kit
1 Kit
Cerebrospinal Fluid, Serum
96 Assays
Videos
Documenti