missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fabp1 (Mouse) ELISA Kit

For quantitative measurement of Fabp1 in serum

Marca:  Abnova™ KA2472.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057305

  • 732.00€ / 1 set


Spedizione stimata
23-10-2020
Aggiungi al carrello

Specifica

Specifica

HRP
Fabp1 (Mouse) ELISA Kit is for the quantitative measurement of Fabp1 in serum.
Colorimetric
96 wells Anti-mouse H-FABP antibody coated wells, Enzyme conjugate reagent (11mL), Reference standard stock (1vial), Diluent (25mL), 20X Wash Buffer (50mL), TMB Reagent (11mL), Stop Solution (11mL)
1 Kit
Store the kit at 4°C.
100μL
Mouse
15.63 to 1000ng/mL
Quantification
Fabp1 (Mouse) ELISA Kit
Serum
96 Assays
Videos
Documenti