missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FIGF (Human) ELISA Kit

Marca:  Abnova™ KA3121.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057855

  • 578.00€ / 1 set


Spedizione stimata
26-10-2020
Aggiungi al carrello

Includes: One 96-well plate pre-coated with anti-human VEGF-D antibody, 96 (8 x 12) wells
Standard (360pg/mL), 0.5mL
Standard diluent buffer, 1.5mL
Wash buffer (30x), Dilution: 1:30, 20mL
Sample diluent buffer, 6mL
HRP conjugated anti-human VEGF-D antibody (RTU), 6mL
Stop solution, 6mL
TMB substrate A, 6mL
TMB substrate B, 6mL
Plate sealer, 2 slices
Hermetic bag, 1 bag

Descrizione

Descrizione

This kit is based on standard sandwich enzyme-linked immune-sorbent assay technology
Specifica

Specifica

10 to 300pg/mL
Human
Colorimetric
96 wells
FIGF (Human) ELISA Kit
Cell Culture Supernatant, Plasma, Serum, Tissue Sample, Urine
Store at 4°C for 6 months.
10μL
HRP
FIGF (Human) ELISA Kit is intended for the quantitative measurement of human FIGF.
Quantification
One 96-well plate pre-coated with anti-human VEGF-D antibody (96 (8x12) wells), Standard (360pg/mL): 0.5mL; Standard diluent buffer (1.5mL), Wash buffer (30x), Dilution: 1:30 (20mL), Sample diluent buffer (6mL), HRP conjugated anti-human VEGF-D antibody (RTU): 6mL, Stop solution (6mL), TMB substrate A (6mL), TMB substrate B (6mL), Plate sealer (2 slices), Hermetic bag (1)
RUO
10μL
96 Assays
Videos
Documenti