missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IgA (Human) ELISA Kit

Marca:  Abnova™ KA2132.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057055

  • 366.00€ / 1 set


Spedizione stimata
26-10-2020
Aggiungi al carrello

Descrizione

Descrizione

The IgA (Human) ELISA Kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgA in serum or plasma of Human’s
Specifica

Specifica

12.5 to 800ng/mL
Human
Colorimetric
96 wells
IgA (Human) ELISA Kit
1 Kit
Serum, Plasma
Store the kit at 4°C.
100μL
HRP
IgA (Human) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of human IgA.
Quantification
Diluent concentrate (50mL), Wash solution concentrate (50mL), Enzyme-Antibody Conjugate Concentrate (150μL), TMB substrate solution (12mL), Stop solution (12mL), Microtiter plate (96 well), Calibrator (Lyophilized): 1 vial
ELISA Kit
RUO
100μL
96 Assays
Videos
Documenti