missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IgM (Dog) ELISA Kit

Enzyme-linked immunoassay for the quantitative determination of IgM in Dog biological samples

Marca:  Abnova™ KA2434.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057275

  • 520.00€ / 1 set


Spedizione stimata
23-10-2020
Aggiungi al carrello

Specifica

Specifica

HRP
IgM (Dog) ELISA Kit is for the quantitative measurement of IgM in plasma and serum.
Colorimetric
96 wells Anti dog IgM-coated wells, HRP Conjugate Reagent (11mL), Reference Standard, 20X wash solution (50mL), 10x Diluent (50mL), TMB reagent (11mL), Stop solution (11mL)
96 Assays
Plasma, Serum
96 Assays
Dog
4.7 to 300 ng/mL
Quantification
IgM (Dog) ELISA Kit
RUO
Store the kit at 4°C.
100μL
Videos
Documenti