missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IgM (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human IgM

Marca:  Abnova™ KA1855.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16056885

  • 418.00€ / 1 set


Spedizione stimata
26-10-2020
Aggiungi al carrello

Specifica

Specifica

Biotin
IgM (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IgM.
Colorimetric
96-well Antibody coated microwell, 200ng Human IgG3 standard, 1 vial Biotinylated IgM Antibody (50X), 30mL MIX diluent concentrate (10X), 30mL x 2 Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromogen Substrate, 12mL stop solution
IgM (Human) ELISA kit
Cell Culture Supernatant, Milk, Plasma, Saliva, Serum, Urine
50μL
Human
1.5ng/mL
Quantification
IgM (Human) ELISA Kit
1 Kit
Store components of the kit at 4°C or -20°C as described in the protocol.
Videos
Documenti