missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL1RN (Human) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Human IL-1RN
dietro preventivo
Specifica
Intervallo di calibrazione | 31.2 to 2000pg/mL |
---|---|
Metodo di rilevamento | Colorimetric |
Includes:
- Lyophilized recombinant Human IL-1RA standard, 10ng/tube x 2
- One 96-well plate precoated with anti-Human IL-1RA antibody, 1 in no.
- Sample diluent buffer, 30mL
- Biotinylated anti- Human IL-1RA antibody, dilution 1:100, 130μLl
- Antibody diluent buffer, 12mL
- Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
- ABC diluent buffer, 12mL
- TMB color developing agent, 10mL
- TMB stop solution, 10mL
Specifica
31.2 to 2000pg/mL | |
Colorimetric |