missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ LGALS3 (Human) ELISA Kit

Marca:  Abnova™ KA3358.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057945

  • 578.00€ / 1 set


Spedizione stimata
27-10-2020
Aggiungi al carrello

Includes: 96-well plate precoated with anti-human Galectin-3 antibody, 1
Lyophilized recombinant human Galectin-3 standard, 10ng/tube × 2
Biotinylated anti-human Galectin-3 antibody (dilution 1:100), 130μL
Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
Sample diluent buffer, 30mL
Antibody diluent buffer, 12mL
ABC diluent buffer, 12mL
TMB color developing agent, 10mL
TMB stop solution, 10mL

Descrizione

Descrizione

LGALS3 (Human) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology
Specifica

Specifica

156 to 10000pg/mL
Human
10pg/mL
Quantification
Antibody coated microwells (96 wells), Lyophilized recombinant Human Galectin-3standard (10ng/tube x 2), Sample diluent buffer (30mL), Biotinylated anti-Human CST3 antibody (130UL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase complex (ABC): 130UL, ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
ELISA Kit
For research use only (RUO)
100μL
Natural and recombinant human Galectin-3
96 Assays
Biotin
LGALS3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human LGALS3.
Colorimetric
96 wells
LGALS3 (Human) ELISA Kit
1 Kit
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
<10pg/mL
Store the kit at-20°C.
100μL
Videos
Documenti