missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NT-proBNP (Human) ELISA Kit

Marca:  Abnova™ KA3099.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057835

  • 578.00€ / 1 set


Spedizione stimata
27-10-2020
Aggiungi al carrello

Includes: One 96-well plate pre-coated with anti-human NT-proBNP antibody, 96 (8x12) wells
Human NT-proBNP standards, 10ng/tube x 2
Sample/Standard diluent buffer, 30mL
HRP conjugated anti-human NT-proBNP antibody (Concentrated): Dilution: 1:100 (store at 4°C, do not store at -20°C),150μL
Antibody diluent buffer, 12mL
TMB substrate, 10mL
Stop solution, 10mL
Wash buffer: (25x): Dilution: 1:25, 30mL

Descrizione

Descrizione

The kit is based on sandwich enzyme-linked immune-sorbent assay technology
Specifica

Specifica

31.2 to 2000pg/mL
Human
2pg/mL
Quantification
One 96-well plate pre-coated with anti-human NT-proBNP antibody (96 (8 x 12) wells, Human NT-proBNP standards: (1μg/mL): 20μL x 2, Sample/Standard diluent buffer (30mL), HRP conjugated anti-human NT-proBNP antibody (Concentrated): Dilution: 1:100 (150μL), Antibody diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer: (25x), Dilution: 1:25 (30mL)
RUO
100μL
Store the kit at 4°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
100μL
HRP
NT-proBNP (Human) ELISA Kit is intended for the quantitative measurement of human NT-proBNP.
Colorimetric
96 wells
NT-proBNP (Human) ELISA Kit
Cell Culture Supernatant, Plasma, Serum
2pg/mL
96 Assays
Videos
Documenti