missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Rubella virus IgG ELISA Kit

solid phase Enzyme Immunoassay for the quantitative measurement of IgG class autoantibodies against Rubella virus.

Marca:  Abnova™ KA1458.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16056655

  • 263.00€ / 1 set


Spedizione stimata
21-10-2020
Aggiungi al carrello

Specifica

Specifica

HRP
Colorimetric
96-well ELISA break away strip coated with specific Antigen, 1.2mL negative Control, 1.2mL/ each vial calibrator (10, 20, 100 and 200 IU), 12mL Antigen/Px Conjugate, 80mL Wash Buffer concentrate, 100mL Dilution Buffer, 13mL Chromogenic Substrate, 15mL stop solution
Rubella Virus IgG ELISA kit
RUO
Store at 4°C on dry atmosphere away from light.
100μL
Rubella virus IgG ELISA Kit is a solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Rubella virus.
Quantification
Rubella virus IgG ELISA Kit
1 Kit
Serum
96 Assays
Videos
Documenti