missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TSLP (Human) ELISA Kit

Marca:  Abnova™ KA2197.1Kit

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057115

  • 578.00€ / 1 set


Spedizione stimata
26-10-2020
Aggiungi al carrello

Includes: Microstrips Coated w/Capture Antibody, 12 x 8 well Microstrips
Protein Standard, Lyophilized (100ng), Red container
Biotinylated Detection Antibody, Lyophilized (1 vial), Yellow container
400x Streptavidin-HRP, 30µL, Blue container
Wash Buffer (10x), 50mL, Clear container
Assay Diluent, 50mL, Clear container
Ready-to-Use Substrate, 12mL, Brown container
Stop Solution, 12mL, Clear container
Adhesive Plate Sealers, 4 Sheets

Descrizione

Descrizione

This immunoassay utilizes the quantitative technique of a “Sandwich” Enzyme-Linked Immunosorbent Assay (ELISA) where the target protein (antigen) is bound in a “sandwich” format by the primary capture antibodies coated to each well-bottom and the secondary detection antibodies added subsequently by the investigator
Specifica

Specifica

Biotin
TSLP (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human TSLP.
Quantification
Antibody coated microwells (96 wells), Biotin-conjugated detection antibody (1 vial), Avidin-HRP conjugate (10UL), Cytokine protein standard (1 vial); Ready-to-use substrate (11mL), Stop solution (11mL), Wash buffer 10X (50mL), Protein standard diluent (15mL), Detection antibody diluent (15mL), HRP diluent (15mL)
ELISA Kit
RUO
100μL
Store the kit at 4°C.
100μL
Human
Colorimetric
96 wells
TSLP (Human) ELISA Kit
1 Kit
Cell Lysate, Plasma, Serum
16-2000pg/mL
96 assays
Videos
Documenti