Learn More
Karl Hecht™ AC-15 Blood Cell Counter
Product Code. 10368861
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10368861 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10368861 Supplier Karl Hecht™ Supplier No. 40345015
  • ONLINE EXCLUSIVE PRICE
    1425.77€
    848.25€ / pezzi 1
    Risparmia 577.52€
    40% Sconto
Spedizione stimata: 09-12-2026
An electronic counter and storage device for the microscopial analysis of differential blood count.
  • For manual differential counting
  • Microscopic image may be monitored continuously while the resulting figures are recorded “blindly”
  • 10 control keys, 15 memory keys with LED display
  • Four defined programs and one free counting program: LEUKO, MYELO, RETI, RBC, FREE
  • Results in absolute values and percentage
  • With cable, safety plug and operating instructions
Specifications
Product Type Blood Cell Counter
Description 230 V, 50 Hz
Memory 15 memory keys
Quantity Each
For Use With (Application) Microscopic analysis of differential blood count
Includes Cable, safety plug, instructions