Description
- Three adjustable flow adapters are available for the C column series (AC 10, AC 16, and AC 26),
- Enabling optimal bed heights for each application, as are thermostatic jackets for precise temperature control
Specifications
Specifications
| Materiale | Gomma al fluoro, gomma fluorocarbonica (FPM), poliammide, polietilene, polipropilene, acciaio inox |
| Tipo | Adattatore di flusso AC 16 |
| Quantità | Confezione da 1 |