Learn More
ACAD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA565455
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Immunogen sequence: DELNEINQFLG PVEKFFTEEV DSRKIDQEGK IPDETLEKLK SLGLFGLQVP EEYGGLGFSN TMYSRLGEII SMDGSITVTL A Highest antigen sequence identity to the following orthologs - mouse 87%, rat 85%.
ACAD9 Polyclonal antibody specifically detects ACAD9 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifica
ACAD9 | |
Polyclonal | |
Unconjugated | |
Acad9 | |
2600017P15Rik; 4732402K02; ACAD9; ACAD-9; Acyl-CoA dehydrogenase family member 9; acyl-CoA dehydrogenase family member 9, mitochondrial; acyl-CoA dehydrogenase family, member 9; acyl-Coenzyme A dehydrogenase family, member 9; C630012L17Rik; Complex I assembly factor ACAD9, mitochondrial; NPD002; Nyggf2; very-long-chain acyl-CoA dehydrogenase VLCAD; very-long-chain acyl-CoA dehydrogenase VLCAD homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28976 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H845 | |
Acad9 | |
Recombinant Human ACAD9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only